L-serine methyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Ser-OMe.HCl

13267-0050 13267-0250 13267-1000
TS13267-0050EA 33 USD
TS13267-0050 TS13267-0250 TS13267-1000
L-serine methyl ester hydrochloride 98%
L-serine methyl ester hydrochloride
Formula: C₄H₉NO₃
MW: 155.58 g/mol
Melting Pt: 160…165 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00063680
CAS Number: 5680-80-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Water ≤1.0 % (K.F.)
Sulfated ash ≤0.1 %
Appearance of solution (5% in methanol) clear colorless to almost colorless
Specific optical rotation +3.5° to +5.5° (20°C, 589 nm) (c=2, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR