L-Selenomethionine 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: L-SelenoMethionine, (S)-2-Amino-4-(methylseleno)butyric Acid, H-Se-Met-OH

25996-0010 25996-0025 25996-2500
TS25996-0010EA 322 USD
TS25996-0010 TS25996-0025 TS25996-2500
L-Selenomethionine 99+%
L-Selenomethionine
Formula: C₅H₁₁NO₂Se
MW: 196.11 g/mol
Melting Pt: 265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037210
CAS Number: 3211-76-5
UN: 3283
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Glistening powder
Infrared spectrum Conforms
HPLC ≥99.0 %
Enantiomeric excess ≥99.0 % (HPLC-CHIR)
Specific optical rotation +17° to +19° (20°C, 589 nm) (c=1, 1 N HCl)
Titration with HClO4 99.0 to 100.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR