L-Histidine methyl ester dihydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: L(+)-Histidine methyl ester dihydrochloride, H-His-OMe.2HCl, L-Methyl histidinate dihydrochloride, H-His-OMe·2HCl

12082-0050 12082-0250 12082-1000
TS12082-0050EA 28 USD
TS12082-0050 TS12082-0250 TS12082-1000
L-Histidine methyl ester dihydrochloride 97%
L-Histidine methyl ester dihydrochloride
Formula: C₇H₁₁N₃O₂
Melting Pt: 207 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012701
CAS Number: 7389-87-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
HPLC ≥96.0 %
Specific optical rotation +8° to +11° (20°C, 589 nm) (c=2, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR