L-Adrenaline bitartrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

43014-0010 43014-0050 43014-0250
TS43014-0010EA 35 USD
TS43014-0010 TS43014-0050 TS43014-0250
L-Adrenaline bitartrate 98+%
L-Adrenaline bitartrate
Formula: C₉H₁₃NO₃
Melting Pt: 155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035077
CAS Number: 51-42-3
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 152 °C (Dec)
Loss on drying ≤0.5 %
Specific optical rotation -20° to -24° (20 °C, 589 nm) (c=4,1M HCl)
Titration with HClO4 ≥98.0 %
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR