Ketoconazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (+/-)-cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine

45547-0010 45547-0050 45547-0250
TS45547-0010EA 79 USD
TS45547-0010 TS45547-0050 TS45547-0250
Ketoconazole 98%
Ketoconazole
Formula: C₂₆H₂₈Cl₂N₄O₄
MW: 531.43 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00058579
CAS Number: 65277-42-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 148 to 152 °C
Loss on drying ≤0.5 % (80 °C, 4 h) (vacuum)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR