Imatinib mesylate 98%

Supplier: Thermo Scientific Chemicals

46108-0010 46108-0050
TS46108-0010EA 251 USD
TS46108-0010 TS46108-0050
Imatinib mesylate 98%
Imatinib mesylate
Formula: C₂₉H₃₁N₇O·CH₄SO₃
Melting Pt: 209 °C
Storage Temperature: Freezer
MDL Number: MFCD04307699
CAS Number: 220127-57-1

Order Now

Specification Test Results

Appearance White to light yelow or light beige Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR