Furosemide 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-(Aminosulphonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 4-Chloro-N-furfuryl-5-sulphamoylanthranilic acid

44897-0050 44897-0010
TS44897-0050EA 190 USD
TS44897-0050 TS44897-0010
Furosemide 97+%
Furosemide
Formula: C₁₂H₁₁ClN₂O₅S
MW: 330.74 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00010549
CAS Number: 54-31-9

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR