Ethyl cellulose, 22 cps 48% (ethoxyl content)
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethylcellulose
19097-0025
19097-2500
19097-5000
TS19097-0025EA
628
USD
TS19097-0025
TS19097-2500
TS19097-5000
Ethyl cellulose, 22 cps 48% (ethoxyl content)
Ethyl cellulose
Storage Temperature:
Ambient |
MDL Number:
MFCD00131037 CAS Number: 9004-57-3 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or granules |
Infrared spectrum | Conforms |
Assay | 48 to 49.5 % (Ethoxyl) |
Viscosity | 18 to 24 cP (5% soln. in 80/20 toluene/ethanol, 25°C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...