D-Valine methyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals

39872-0010 39872-0050
TS39872-0010EA 38 USD
TS39872-0010 TS39872-0050
D-Valine methyl ester hydrochloride 99%
D-Valine methyl ester hydrochloride
Formula: C₆H₁₃NO₂
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00237309
CAS Number: 7146-15-8

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Specific optical rotation -13.5° to -17.5° (20°C, 589 nm) (c=2,H2O)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR