D-Phenylalanine methyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-Methyl 2-amino-3-phenylpropanoate hydrochloride, H-D-Phe-OMe.HCl, H-D-Phe-OMe·HCl
44262-0050
44262-0250
TS44262-0050EA
50
USD
TS44262-0050
TS44262-0250
D-Phenylalanine methyl ester hydrochloride 98%
D-Phenylalanine methyl ester hydrochloride
Formula:
C₁₀H₁₃NO₂ Melting Pt: 157…159 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066112 CAS Number: 13033-84-6 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Specific optical rotation | -32° to -38° (20°C, 589 nm) (c=1 in EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...