Doxycycline hyclate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Doxycycline Hydrochloride Hemiethanolate Hemihydrate
44606-0050
44606-1000
44606-0250
TS44606-0050EA
50
USD
TS44606-0050
TS44606-1000
TS44606-0250
Doxycycline hyclate 97%
Doxycycline hyclate
Formula:
C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O MW: 512.94 g/mol (503.94 anhy) Storage Temperature: Refrigerator |
MDL Number:
MFCD07357237 CAS Number: 24390-14-5 |
Specification Test Results
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Water | 1.4 to 2.8 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...