DL-Homocysteinethiolactone hydrochloride 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
DL-3-Aminotetrahydrothiophen-2-one Hydrochloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone HCl
12083-0250
12083-1000
TS12083-0250EA
38
USD
TS12083-0250
TS12083-1000
DL-Homocysteinethiolactone hydrochloride 99%
DL-Homocysteinethiolactone hydrochloride
Formula:
C₄H₇NOS MW: 153.62 g/mol Melting Pt: 199…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012724 CAS Number: 6038-19-3 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...