DL-Alanine methyl ester hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-DL-Ala-OMe.HCl, H-DL-Ala-OMe·HCl

39331-0010 39331-0050 39331-0250 39331-1000
TS39331-0010EA 25 USD
TS39331-0010 TS39331-0050 TS39331-0250 TS39331-1000
DL-Alanine methyl ester hydrochloride 98+%
DL-Alanine methyl ester hydrochloride
Formula: C₄H₉NO₂
Melting Pt: 157…158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00035523
CAS Number: 13515-97-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR