Dimethyl-L-glutamate hydrochloride 99%, pure

Supplier: Thermo Scientific Chemicals

Synonyms: L-Glutamic Acid Dimethyl Ester Hydrochloride, H-Glu(OMe)-OMe.HCl

44194-0100 44194-0500
TS44194-0100EA 63 USD
TS44194-0100 TS44194-0500
Dimethyl-L-glutamate hydrochloride 99%, pure
Dimethyl-L-glutamate hydrochloride
Formula: C₇H₁₃NO₄
MW: 211.64 g/mol
Melting Pt: 91…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038879
CAS Number: 23150-65-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Specific optical rotation +25° to +27° (20°C, 589 nm) (c=5 in H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR