Dimethyl-L-aspartate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-Aminosuccinic Acid Dimethyl Ester Hydrochloride, L-Asparaginic Acid Dimethyl Ester Hydrochloride, Dimethyl (S)-Aminosuccinate Hydrochloride, H-Asp(ome)-OMe HCl, Dimethyl L-Asparaginate Hydrochloride, H-Asp(OMe)-OMe.HCl, L-Aspartic Acid Dimethyl Ester Hydrochloride
37666-0050
37666-0250
TS37666-0050EA
69
USD
TS37666-0050
TS37666-0250
Dimethyl-L-aspartate hydrochloride 98%
Dimethyl-L-aspartate hydrochloride
Formula:
C₆H₁₁NO₄ MW: 197.62 g/mol Melting Pt: 115…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038878 CAS Number: 32213-95-9 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Specific optical rotation | +13° to +17° (20°C, 589 nm) (c=1, CH3OH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...