Coenzyme Q10 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (all-E)-2-(3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, Ubiquinone 10, Ubidecarenone

45795-0010 45795-0050 45795-0250
TS45795-0010EA 63 USD
TS45795-0010 TS45795-0050 TS45795-0250
Coenzyme Q10 98%
Coenzyme Q10
Formula: C₅₉H₉₀O₄
MW: 863.37 g/mol
Melting Pt: 49 °C
Storage Temperature: Freezer
MDL Number: MFCD00042919
CAS Number: 303-98-0

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific absorbance A (1%/1cm) ≥162 (275 nm, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR