Clofazimine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine, N,5-Bis(4-chlorophenyl)-3-(isopropylimino)-3,5-dihydrophenazin-2-amine

46176-0010 46176-0050
TS46176-0010EA 66 USD
TS46176-0010 TS46176-0050
Clofazimine 98%
Clofazimine
Formula: C₂₇H₂₂Cl₂N₄
MW: 473.40 g/mol
Melting Pt: 219 °C
Storage Temperature: Ambient
MDL Number: MFCD00056793
CAS Number: 2030-63-9

Order Now

Specification Test Results

Appearance Orange to red to brown Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C)
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR