Citalopram hydrobromide (by HPLC)

Supplier: Thermo Scientific Chemicals
Warning

46231-0010 46231-0050
TS46231-0010EA 82.81 USD
TS46231-0010 TS46231-0050
Citalopram hydrobromide (by HPLC)
Citalopram hydrobromide
Formula: C₂₀H₂₁FN₂O
Melting Pt: 185…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02101306
CAS Number: 59729-32-7

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 4 h)
Solubility (1 % in ethanol) Clear colorless
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR