Ciprofloxacin 98%

Supplier: Thermo Scientific Chemicals

44962-0050 44962-0250
TS44962-0050EA 58 USD
TS44962-0050 TS44962-0250
Ciprofloxacin 98%
Ciprofloxacin
Formula: C₁₇H₁₈FN₃O₃
Storage Temperature: Freezer
MDL Number: MFCD00185755
CAS Number: 85721-33-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Sulfated ash ≤0.1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR