Chloroquine diphosphate salt 98%

Supplier: Thermo Scientific Chemicals
Warning

45524-0250 45524-1000 45524-5000
TS45524-0250EA 62 USD
TS45524-0250 TS45524-1000 TS45524-5000
Chloroquine diphosphate salt 98%
Chloroquine diphosphate salt
Formula: C₁₈H₂₆ClN₃
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00069852
CAS Number: 50-63-5

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (105°C, 16 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR