Ceftriaxone disodium salt hemiheptahydrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ceftriaxone sodium hydrate, Ceftriaxone sodium trihydrate, Ceftriaxone sodium salt hydrate, (6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-3-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, disodium salt, hemiheptahydrate

45542-0250 45542-0010 45542-0050
TS45542-0250EA 251 USD
TS45542-0250 TS45542-0010 TS45542-0050
Ceftriaxone disodium salt hemiheptahydrate
Ceftriaxone disodium salt hemiheptahydrate
Formula: C₁₈H₁₈N₈O₇S₃
Melting Pt: 236 °C
Storage Temperature: Ambient
MDL Number: MFCD01750405
CAS Number: 104376-79-6

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤10 %
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR