Cefotaxime sodium salt 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefotaxime sodium

45495-0010 45495-0050 45495-0250
TS45495-0010EA 79 USD
TS45495-0010 TS45495-0050 TS45495-0250
Cefotaxime sodium salt 95%
Cefotaxime sodium salt
Formula: C₁₆H₁₇N₅O₇S₂
MW: 477.44 g/mol
Melting Pt: 201…210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079073
CAS Number: 64485-93-4

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤3.0 % (105°C, 8 hrs)
HPLC ≥94 %
pH 4.5 to 6.5
Specific optical rotation +58° to +65° (20°C, 589 nm) (c=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR