Cefazolin sodium salt 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium (6R,7R)-7-(2-(1H-tetrazol-1-yl)acetamido)-3-(((5-methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

45521-0010 45521-0050
TS45521-0010EA 153 USD
TS45521-0010 TS45521-0050
Cefazolin sodium salt 98%
Cefazolin sodium salt
Formula: C₁₄H₁₄N₈NaO₄S₃
MW: 476.48 g/mol
Melting Pt: 170…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056883
CAS Number: 27164-46-1

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 170 to 177 °C
Water ≤6 %
HPLC ≥97.5 %
Specific optical rotation -24° to -10° (20 °C, 589 nm) (c=1,H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR