β-methasone 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 9ɑ-Fluoro-11β,17ɑ,21-trihydroxy-16β-methyl-1,4-pregnadiene-3,20-dione

45907-0010 45907-0050 45907-0250
TS45907-0010EA 45 USD
TS45907-0010 TS45907-0050 TS45907-0250
β-methasone 97%
Betamethasone
Formula: C₂₂H₂₉FO₅
MW: 392.47 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00062969
CAS Number: 378-44-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 235 to 245 °C
Loss on drying ≤0.5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR