Balsalazide disodium salt dihydrate

Supplier: Thermo Scientific Chemicals

46509-0050 465090010
TS46509-0050EA 180 USD
TS46509-0050 TS465090010
Balsalazide disodium salt dihydrate
Balsalazide disodium salt dihydrate
Formula: C₁₇H₁₇N₃Na₂O₈
Storage Temperature: Refrigerator
MDL Number: MFCD01631117
CAS Number: 150399-21-6

Order Now

Specification Test Results

Appearance Yellow to yellow-red Crystalline powder
Infrared spectrum Conforms
Water 7 to 10 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR