Atracurium besylate (mixture of isomers) 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Atracurium Besylate, 51W89, BW-33A, 2,2'-[1,5-Pentanediylbis[oxy(3-oxo-3,1-propanediyl)]]bis[1-(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methylisoquinolinium] Dibenzenesulfonate (mixture of isomers), 2,2'-[1,5-Pentanediylbis[oxy(3-oxo-3,1-propanediyl)]]bis[1-(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methylisoquinolinium] Dibenzenesulphonate, Tracrium

46147-0010 46147-0050
TS46147-0010EA 105 USD
TS46147-0010 TS46147-0050
Atracurium besylate (mixture of isomers) 96%
Atracurium besylate (mixture of isomers)
Formula: C₆₅H₈₂N₂O₁₈S₂
MW: 1243.49 g/mol
Melting Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00797403
CAS Number: 64228-81-5

Order Now

Specification Test Results

Appearance White to yellowish-white Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥95.0 % (mix of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR