Atorvastatin calcium salt trihydrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Calcium (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate trihydrate, Atorvastatin Calcium Salt Trihydrate, Atorvastatin hemicalcium salt sesquihydrate, Atorvastatin calcium

46481-0010 46481-0050
TS46481-0010EA 95 USD
TS46481-0010 TS46481-0050
Atorvastatin calcium salt trihydrate
Atorvastatin calcium salt trihydrate
Formula: C₆₆H₇₄CaF₂N₄O₁₃
MW: 1209.39 g/mol
Melting Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD09752074
CAS Number: 344423-98-9

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water 3.5 to 5.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR