Adapalene

Supplier: Thermo Scientific Chemicals
Warning

46450-0010 464500050
TS46450-0010EA 131 USD
TS46450-0010 TS464500050
Adapalene
Adapalene
Formula: C₂₈H₂₈O₃
Melting Pt: 324 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03106112
CAS Number: 106685-40-9

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C, 4 h)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR