5-Aza-2'-deoxycytidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one

39681-0500 39681-0100
TS39681-0500EA 466 USD
TS39681-0500 TS39681-0100
5-Aza-2'-deoxycytidine 98%
5-Aza-2'-deoxycytidine
Formula: C₈H₁₂N₄O₄
MW: 228.21 g/mol
Melting Pt: 200 °C
Storage Temperature: Freezer
MDL Number: MFCD00043011
CAS Number: 2353-33-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR