4-Benzyl-L-aspartate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Aspartic acid beta-benzyl ester, β-Benzyl L-aspartate, H-Asp(obzl)-OH, beta-Benzyl L-aspartate, L-Aspartic acid 4-benzyl ester, L-Aspartic Acid 4-Benzyl Ester, beta-Benzyl L-Aspartate, H-Asp(OBzl)-OH, 4-Benzyl L-Aspartate, H-Asp(OBn)-OH, L-Aspartic Acid beta-Benzyl Ester, L-Aspartic acid β-benzyl ester

44193-0050 44193-0250
TS44193-0050EA 78 USD
TS44193-0050 TS44193-0250
4-Benzyl-L-aspartate 98%
4-Benzyl-L-aspartate
Formula: C₁₁H₁₃NO₄
Melting Pt: 207…208 °C
Storage Temperature: Freezer
MDL Number: MFCD00037208
CAS Number: 2177-63-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +26° to +30° (20°C, 589 nm) (c=1 in dil HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR