4,4,4,4',4',4'-Hexafluoro-DL-valine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44654-0010
44654-0050
TS44654-0010EA
247
USD
TS44654-0010
TS44654-0050
4,4,4,4',4',4'-Hexafluoro-DL-valine 97%
4,4,4,4',4',4'-Hexafluoro-DL-valine
Formula:
C₅H₅F₆NO₂ Melting Pt: 207…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014349 CAS Number: 16063-80-2 |
Specification Test Results
Appearance | White to off-white Powder or solid |
Infrared spectrum | Conforms |
Assay | 96 to 104 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...