3-(Methylamino)propionitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(N-Methylamino)propionitrile, N-(2-Cyanoethyl)methylamine, N-Methyl-β-alaninenitrile, 3-(Methylamino)propanenitrile

12622-0050 12622-0500
TS12622-0050EA 36 USD
TS12622-0050 TS12622-0500
3-(Methylamino)propionitrile 98%
3-(Methylamino)propionitrile
Formula: C₄H₈N₂
MW: 84.12 g/mol
Boiling Pt: 182…186 °C (760 mmHg)
Density: 0.89 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001954
CAS Number: 693-05-0

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4320 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %
UV at 310 nm A: ≤0.046
Non-volatile matter ≤0.0003 %
Acidity ≤0.0005 meq/g
Alkalinity ≤0.003 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR