2-Methyl-5-nitroimidazole-1-ethanol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Metronidazole

21034-0050 21034-1000 21034-5000
TS21034-0050EA 90 USD
TS21034-0050 TS21034-1000 TS21034-5000
2-Methyl-5-nitroimidazole-1-ethanol 99%
2-Methyl-5-nitroimidazole-1-ethanol
Formula: C₆H₉N₃O₃
MW: 171.16 g/mol
Melting Pt: 159…163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009750
CAS Number: 443-48-1

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 163 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR