2-Fluoroadenine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Amino-2-fluoropurine, 6-Amino-2-fluoro-7H-purine, 2-Fluoro-7H-purin-6-amine

45916-0010 45916-0050
TS45916-0010EA 182 USD
TS45916-0010 TS45916-0050
2-Fluoroadenine 97%
2-Fluoroadenine
Formula: C₅H₄FN₅
MW: 153.12 g/mol
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD01632749
CAS Number: 700-49-2

Order Now

Specification Test Results

Appearance White to brown Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR