2-Deoxy-D-galactose 99%

Supplier: Thermo Scientific Chemicals

11197-0010 11197-0050
TS11197-0010EA 104 USD
TS11197-0010 TS11197-0050
2-Deoxy-D-galactose 99%
2-Deoxy-D-galactose
Formula: C₆H₁₂O₅
MW: 164.16 g/mol
Melting Pt: 109…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00014649
CAS Number: 1949-89-9

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥98.5 % (ELSD)
Specific optical rotation +56° to +60° (20°C, 589 nm) (c=2, H2O, 5 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR