2'-Deoxy-2'-fluorouridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2'-Fluoro-2'-deoxyuridine, 1-((2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

45500-0010 45500-0250
TS45500-0010EA 48 USD
TS45500-0010 TS45500-0050 TS45500-0250
2'-Deoxy-2'-fluorouridine 97%
2'-Deoxy-2'-fluorouridine
Formula: C₉H₁₁FN₂O₅
MW: 246.19 g/mol
Melting Pt: 151 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01317293
CAS Number: 784-71-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 153 °C
Loss on drying ≤1 % (1 g, 105 °C, 3 h)
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR