2-(2-Chlorophenyl)glycine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-DL-Phg(2-Cl)-OH, 2-Amino-2-(2-chlorophenyl)acetic Acid, (±)-2-Chlorophenylglycine

43334-0250 43334-1000
TS43334-0250EA 102 USD
TS43334-0250 TS43334-1000
2-(2-Chlorophenyl)glycine 98%
2-(2-Chlorophenyl)glycine
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Melting Pt: 182.9…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00049324
CAS Number: 88744-36-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR