1,6-Anhydro-β-D-glucose 99+%
Supplier: Thermo Scientific Chemicals
27095-0010
27095-0050
TS27095-0010EA
91
USD
TS27095-0010
TS27095-0050
1,6-Anhydro-β-D-glucose 99+%
1,6-Anhydro-beta-D-glucose
Formula:
C₆H₁₀O₅ Melting Pt: 170…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063248 CAS Number: 498-07-7 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Water | ≤1.0 % (K.F.) |
Solubility | (5 % in water) Clear colorless |
HPLC | ≥99.0 % (ELSD) |
Specific optical rotation | -64° to -68° (20°C, 589 nm) (c=2, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...