1,2-O-Isopropylidene-ɑ-D-xylofuranose 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,2-O-Isopropylidene-α-D-xylofuranose, (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol, (3aR,5R,6S,6aR)-5-(Hydroxymethyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol

34888-0100 34888-0250 34888-1000
TS34888-0100EA 113 USD
TS34888-0100 TS34888-0250 TS34888-1000
1,2-O-Isopropylidene-ɑ-D-xylofuranose 99%
1,2-O-Isopropylidene-alpha-D-xylofuranose
Formula: C₈H₁₄O₅
MW: 190.20 g/mol
Boiling Pt: 112…114 °C (.1 mmHg)
Melting Pt: 68…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063295
CAS Number: 20031-21-4

Order Now

Specification Test Results

Appearance White Waxy solid or coarse crystals and/or chunks
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation -17° to -19° (20°C, 589 nm) (c=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR