1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

12331-0010 12331-0050 12331-1000
TS12331-0010EA 217 USD
TS12331-0010 TS12331-0050 TS12331-1000
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid
Formula: C₂₂H₂₄N₂O₁₀
Melting Pt: 177…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036255
CAS Number: 85233-19-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR