[(tert-Butoxycarbonyl)aminooxy]acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Boc-(carboxymethoxy)amine, Boc-Aoa-OH, (Boc-aminooxy)acetic Acid

42969-0010 42969-0050
TS42969-0010EA 68 USD
TS42969-0010 TS42969-0050
[(tert-Butoxycarbonyl)aminooxy]acetic acid 98%
[(tert-Butoxycarbonyl)aminooxy]acetic acid
Formula: C₇H₁₃NO₅
MW: 191.18 g/mol
Melting Pt: 111.5…112 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01632027
CAS Number: 42989-85-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 113 °C (Dec)
Titration with HClO4 ≥97.5 %
Loss on ignition 24.5 % to 26.5 % (800 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR