(S)-(+)-Clopidogrel sulfate 98%

Supplier: Thermo Scientific Chemicals

46103-0010 46103-0050
TS46103-0010EA 118 USD
TS46103-0010 TS46103-0050
(S)-(+)-Clopidogrel sulfate 98%
(S)-(+)-Clopidogrel sulfate
Formula: C₁₆H₁₆ClNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00876395
CAS Number: 120202-66-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Specific optical rotation +54° to +58° (20°C, 589 nm) (1 % in Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR