(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 95%, 98% ee

Supplier: Thermo Scientific Chemicals

Synonyms: (1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, Boc-D-AcPAC, (1S,3R)-3-(Boc-amino)cyclopentanecarboxylic Acid, (1S,3R)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

36183-0010 36183-2500
TS36183-0010EA 342 USD
TS36183-0010 TS36183-2500
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 95%, 98% ee
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Melting Pt: 111.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01320857
CAS Number: 261165-05-3

Order Now

Specification Test Results

Appearance White Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥94 % (ELSD)
Enantiomeric excess ≥97.5 % (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR