(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Boc-L-serine benzyl ester, Boc-Ser-OBzl, BOC-L-Serine benzyl ester

43484-0010 43484-0050
TS43484-0010EA 59 USD
TS43484-0010 TS43484-0050
(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 98%
(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate
Formula: C₁₅H₂₁NO₅
MW: 295.3 g/mol
Melting Pt: 67…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00076983
CAS Number: 59524-02-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 65 to 75 °C
HPLC ≥97.5 %
Specific optical rotation -14° to -18° (20 °C, 589 nm) (c=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR