β-D-Ribofuranose-1,2,3,5-tetraacetate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Tetra-O-acetyl-β-D-ribofuranose, (2S,3R,4R,5R)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate

17012-0250 17012-1000
TS17012-0250EA 120 USD
TS17012-0250 TS17012-1000
β-D-Ribofuranose-1,2,3,5-tetraacetate 98+%
beta-D-Ribofuranose-1,2,3,5-tetraacetate
Formula: C₁₃H₁₈O₉
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00005358
CAS Number: 13035-61-5

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (60°C, 2 h) (vacuum)
HPLC ≥98.0 %
Residue after ignition ≤0.1 %
Specific optical rotation -12° to -14° (20°C, 589 nm) (c=10, CHCl3)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR