(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-β-iodo-Ala-OMe, BOC-3-iodo-L-alanine methyl ester, N-Boc-3-iodo-L-alanine Methyl Ester, Boc-β-Iodo-Ala-OMe, Boc-Ala(3-I)-OMe

43846-0010 43846-0050
TS43846-0010EA 138 USD
TS43846-0010 TS43846-0050
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 97%
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate
Formula: C₉H₁₆INO₄
MW: 329.13 g/mol
Melting Pt: 49…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216579
CAS Number: 93267-04-0

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 49 to 53 °C
HPLC ≥96.0 %
Specific optical rotation -3.0° to -5.0° (20 °C, 589 nm) (c=2 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR