Methyl L-tryptophanate hydrochloride 98%

Supplier: Thermo Scientific Chemicals

37665-0050 37665-0250
TS37665-0050EA 36 USD
TS37665-0050 TS37665-0250
Methyl L-tryptophanate hydrochloride 98%
Methyl L-tryptophanate hydrochloride
Formula: C₁₂H₁₄N₂O₂·HCl
Melting Pt: 218…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066134
CAS Number: 7524-52-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Specific optical rotation +17° to +19° (20°C, 589 nm) (c=5, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR