Ethyl-3-aminopropanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: beta-Alanine ethyl ester hydrochloride, H-beta-Ala-OEt.HCl, Ethyl 3-Aminopropionate Hydrochloride, β-Alanine ethyl ester hydrochloride, 3-Aminopropionic Acid Ethyl Ester Hydrochloride, Ethyl 3-aminopropanoate hydrochloride, 3-Aminopropionic acid Ethyl Ester Hydrochloride, H-β-Ala-OEt·HCl, Ethyl 3-aminopropionate HCl

30306-0100 30306-0500
TS30306-0100EA 66 USD
TS30306-0100 TS30306-0500
Ethyl-3-aminopropanoate hydrochloride 98%
Ethyl-3-aminopropanoate hydrochloride
Formula: C₅H₁₁NO₂·HCl
MW: 153.6 g/mol
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00012909
CAS Number: 4244-84-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
TLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR