1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Allopurinol

12183-0050 12183-0250 12183-1000
TS12183-0050EA 54 USD
TS12183-0050 TS12183-0250 TS12183-1000
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one
Formula: C₅H₄N₄O
MW: 136.11 g/mol
Melting Pt: 384 °C
Storage Temperature: Ambient
MDL Number: MFCD00599413
CAS Number: 315-30-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Titration with NaOH ≥97.5 % (on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR