Ritalinic acid

Supplier: Thermo Scientific Chemicals
Warning

46300-0010 463000050
TS46300-0010EA 70 USD
TS46300-0010 TS463000050
Ritalinic acid
Ritalinic acid
Formula: C13H17NO2
Melting Pt: 245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06200695
CAS Number: 19395-41-6

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR